Learn More
Biomatik Corporation Mouse neurotensin (NTS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37483
Encompass_Preferred
Mouse Neurotensin (Nts) Elisa Kit for detecting Neurotensin in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9D3P9; Nts. Target Synonyms: Nts; Neurotensin/neuromedin N [Cleaved into: Large neuromedin N; NmN-125); Neuromedin N; NN; NmN); Neurotensin; NT); Tail peptide]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8606