Learn More
Biomatik Corporation Mouse pancreatic alpha amylase (AMY2A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37542
Encompass_Preferred
Mouse Pancreatic Alpha Amylase (Amy2A) Elisa Kit for detecting Pancreatic Alpha Amylase (Amy2A) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 9.38-600mU/ml. Sensitivity: 2.3mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P00688; Amy2. Target Synonyms: Amy2; Amy2a; Pancreatic alpha-amylase; PA; EC 3.2.1.1; 1; 4-alpha-D-glucan glucanohydrolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8621