Learn More
Biomatik Corporation Mouse plasminogen activator inhibitor 1, PAI-1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37588
Encompass_Preferred
Mouse Plasminogen Activator Inhibitor 1, Pai-1 Elisa Kit for detecting Serpin Peptidase Inhibitor, Clade E (Nexin, Plasminogen Activator Inhibitor Type 1), Member 1 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.9-60ng/ml. Sensitivity: 0.22ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P22777; Serpine1. Target Synonyms: Serpine1; Mr1; Pai1; Planh1; Plasminogen activator inhibitor 1; PAI; PAI-1; Endothelial plasminogen activator inhibitor; Serpin E1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8632