Learn More
Biomatik Corporation Mouse thrombospondin 1, TSP-1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37914
Encompass_Preferred
Mouse Thrombospondin 1, Tsp-1 Elisa Kit for detecting Thrombospondin 1, Tsp-1 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.27-200ng/ml. Sensitivity: 0.27ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P35441; Thbs1. Target Synonyms: Thbs1; Tsp1; Thrombospondin-1; Glycoprotein G. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8694