Learn More
Biomatik Corporation Mouse Vascular Endothelial cell Growth Factor C, VEGF-C ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38027
Encompass_Preferred
Mouse Vascular Endothelial Cell Growth Factor C, Vegf-C Elisa Kit for detecting Vascular Endothelial Growth Factor C in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P97953; Vegfc. Target Synonyms: VegfcVascular endothelial growth factor C; VEGF-C; Flt4 ligand; Flt4-L; Vascular endothelial growth factor-related protein; VRP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8711