Learn More
Biomatik Corporation Mouse very low density lipoprotein (VLDL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38038
Encompass_Preferred
Mouse Very Low Density Lipoprotein (Vldl) Elisa Kit for detecting Very Low Density Lipoprotein(Vldl) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8712