Learn More
Biomatik Corporation Pig fibroblast growth factor 19 (FGF19) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38180
Encompass_Preferred
Pig Fibroblast Growth Factor 19 (Fgf19) Elisa Kit for detecting Fibroblast Growth Factor 19(Fgf19) in serum, plasma. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: F1RY79. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8735