Learn More
Biomatik Corporation Pig inducible nitric oxide synthase (iNOS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38219
Encompass_Preferred
Pig Inducible Nitric Oxide Synthase (Inos) Elisa Kit for detecting Inducible Nitric Oxide Synthase(Inos) in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.05-40U/ml. Sensitivity: 0.05u/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P29477; Nos2. Target Synonyms: Nos2; InoslNitric oxide synthase; inducible; EC 1.14.13.39; Inducible NO synthase; Inducible NOS; iNOS; Macrophage NOS; MAC-NOS; NOS type II; Peptidyl-cysteine S-nitrosylase NOS2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8743