Learn More
Biomatik Corporation Pig N-terminal pro-brain natriuretic peptide (NT-proBNP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38293
Encompass_Preferred
Pig N-Terminal Pro-Brain Natriuretic Peptide (Nt-Probnp) Elisa Kit for detecting N-Terminal Pro-Brain Natriuretic Peptide (Nt-Probnp) in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8762