Learn More
Biomatik Corporation Rabbit Tumor necrosis factor , TNF ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38559
Encompass_Preferred
Rabbit Tumor Necrosis Factor Α, Tnfα Elisa Kit for detecting Tumor Necrosis Factor (Tnf Superfamily, Member 2) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04924; TNF. Target Synonyms: TNF; TNFA; TNFSF2; Tumor necrosis factor; Cachectin; TNF-alpha; Tumor necrosis factor ligand superfamily member 2; TNF-a. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8796