Learn More
Biomatik Corporation Rat amyloid beta peptide 1-40, A1-40 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38654
Encompass_Preferred
Rat Amyloid Beta Peptide 1-40, Aβ1-40 Elisa Kit for detecting Amyloid Beta Peptide 1-40, Aβ1-40 in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40pg/ml. Sensitivity: 0.156pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: amyloid beta peptide 1-40; Aβ1-40; amyloid β peptide 1-40. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8811