Learn More
Biomatik Corporation Rat angiotensin II (ANG-) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38669
Encompass_Preferred
Rat Angiotensin Ii (Ang-Ii) Elisa Kit for detecting Angiotensin Ii in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 4.7-300pg/ml. Sensitivity: 1.17pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: angiotensin II; ANG-II; angiotensin 2; ANG-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8816