Learn More
Biomatik Corporation Rat coagulation factor , F ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC38854
Encompass_Preferred
Rat Coagulation Factor Ⅹ, FⅩ Elisa Kit for detecting Coagulation Factor X in serum, plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q63207; F10. Target Synonyms: F10Coagulation factor X; EC 3.4.21.6; Stuart factor) [Cleaved into: Factor X light chain; Factor X heavy chain; Activated factor Xa heavy chain]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8841