Learn More
Biomatik Corporation Rat Glycated hemoglobin A1c, GHbA1c ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39123
Encompass_Preferred
Rat Glycated Hemoglobin A1C, Ghba1C Elisa Kit for detecting Glycated Hemoglobin A1C, Ghba1C in lysates for rbc. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.019-80ng/ml. Sensitivity: 0.019ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8884