Learn More
Biomatik Corporation Rat hepatic lipase, HL ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39183
Encompass_Preferred
Rat Hepatic Lipase, Hl Elisa Kit for detecting Lipase, Hepatic in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P07867; Lipc. Target Synonyms: LipcHepatic triacylglycerol lipase; HL; Hepatic lipase; EC 3.1.1.3; Lipase member C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8891