Learn More
Biomatik Corporation Rat IL-1 Receptor Like 1 (IL1RL1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39211
Encompass_Preferred
Rat Il-1 Receptor Like 1 (Il1Rl1) Elisa Kit for detecting Interleukin 1 Receptor-Like 1 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q62611; Il1rl1. Target Synonyms: Il1rl1; Fit1; Interleukin-1 receptor-like 1; EC 3.2.2.6; Fit-1; Fos-responsive gene 1 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8896