Learn More
Biomatik Corporation Rat Insulin-like growth factor 2, IGF-2 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39231
Encompass_Preferred
Rat Insulin-Like Growth Factor 2, Igf-2 Elisa Kit for detecting Insulin-Like Growth Factor 2 (Somatomedin A) in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01346; Igf2. Target Synonyms: Igf2; Igf-2; Insulin-like growth factor II; IGF-II; Multiplication-stimulating activity; MSA; Multiplication-stimulating polypeptide) [Cleaved into: Insulin-like growth factor II; Preptin]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8903