Learn More
Biomatik Corporation Rat neutrophil elastase, NE ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39487
Encompass_Preferred
Rat Neutrophil Elastase, Ne Elisa Kit for detecting Neutrophil Elastase, Ne in serum, plasma, cell culture supernates, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.5-800ng/ml. Sensitivity: 3.12ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P08246; ELANE. Target Synonyms: Bone marrow serine protease; ELA2; ELANE; Elastase 2; Elastase 2 neutrophil; Elastase neutrophil expressed; Elastase-2; ELNE_HUMAN; GE; Granulocyte derived elastase; HLE; HNE; Human leukocyte elastase; Leukocyte elastase; Medullasin; NE; Neutrophil elastase; PMN E; PMN elastase; Polymorphonuclear elastase; SCN1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8946