Learn More
Biomatik Corporation Rat soluble E-selectin, sE-selectin ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC39704
Encompass_Preferred
Rat Soluble E-Selectin, Se-Selectin Elisa Kit for detecting Selectin E in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P98105; Sele. Target Synonyms: Sele; Elam-1; E-selectin; CD62 antigen-like family member E; Endothelial leukocyte adhesion molecule 1; ELAM-1; Leukocyte-endothelial cell adhesion molecule 2; LECAM2; CD antigen CD62E. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8986