Learn More
Biomatik Corporation Human Factor-related Apoptosis ligand, FASL ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40040
Encompass_Preferred
The Human Factor-related Apoptosis ligand, FASL ELISA Kit allows for quantitative determination of Fas ligand (TNF superfamily, member 6) in serum, plasma, cell culture supernates. Species Reactivity: Human. Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.81 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P48023. Restriction: For Research Use Only.
Catalog No. 50-149-9029