Learn More
Biomatik Corporation Rat intercellular adhesion molecule 1, ICAM-1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40055
Encompass_Preferred
Rat Intercellular Adhesion Molecule 1, Icam-1 Elisa Kit for detecting Intercellular Adhesion Molecule 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 24.321pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q00238; Icam1. Target Synonyms: Icam1; Icam-1; Intercellular adhesion molecule 1; ICAM-1; CD antigen CD54. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9036