Learn More
Biomatik Corporation Mouse thyroxine, T4 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40187
Encompass_Preferred
Mouse Thyroxine, T4 Elisa Kit for detecting Thyroxine, T4 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 20-320ng/ml. Sensitivity: 20ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9082