Learn More
Biomatik Corporation Pig Interferon , IFN- ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40219
Encompass_Preferred
Pig Interferon Γ , Ifn-Γ Elisa Kit for detecting Interferon, Gamma in serum, plasma, tissue homogenates. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 15.6-1000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P17803; IFNG. Target Synonyms: IFNGInterferon gamma; IFN-gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9101