Learn More
Biomatik Corporation Human adrenocorticotropic hormone (ACTH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40235
Encompass_Preferred
The Human adrenocorticotropic hormone (ACTH) ELISA Kit allows for quantitative determination of adrenocorticotropic hormone (ACTH) in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 12.5 pg/mL-200 pg/mL. Sensitivity: 12.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-9110