Learn More
Biomatik Corporation Rat tissue inhibitors of metalloproteinase 2, TIMP-2 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40315
Encompass_Preferred
Rat Tissue Inhibitors Of Metalloproteinase 2, Timp-2 Elisa Kit for detecting Timp Metallopeptidase Inhibitor 2 in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.25-80ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P30121; Timp2. Target Synonyms: Timp2; Timp-2; Metalloproteinase inhibitor 2; Tissue inhibitor of metalloproteinases 2; TIMP-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9147