Learn More
Biomatik Corporation Rat Dihydrotestosterone, DHT ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40341
Encompass_Preferred
Rat Dihydrotestosterone, Dht Elisa Kit for detecting Dihydrotestosterone, Dht in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 10-2000pg/ml. Sensitivity: 5pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9158