Learn More
Biomatik Corporation Human Heat Shock Protein glycoprotein 96, HSP gp96 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40470
Encompass_Preferred
The Human Heat Shock Protein glycoprotein 96, HSP gp96 ELISA Kit allows for quantitative determination of Heat Shock Protein glycoprotein 96, HSP gp96 in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 0.625 ng/mL-40 ng/mL. Sensitivity: 0.156 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-9225