Learn More
Biomatik Corporation Rat Interferon , IFN- ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40490
Encompass_Preferred
Rat Interferon Α, Ifn-Α Elisa Kit for detecting Interferon Α, Ifn-Α in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P05011; Ifna1. Target Synonyms: Ifna1; Interferon alpha-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9235