Learn More
Biomatik Corporation Mouse parathyroid hormone (PTH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40498
Encompass_Preferred
Mouse Parathyroid Hormone (Pth) Elisa Kit for detecting Parathyroid Hormone (Pth) in serum, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9239