Learn More
Biomatik Corporation Human tyrosine hydroxylase, TH ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40673
Encompass_Preferred
Human Tyrosine Hydroxylase, Th Elisa Kit for detecting Tyrosine Hydroxylase in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P07101; TH. Target Synonyms: Dystonia 14; DYT14; DYT5b; EC 1.14.16.2; OTTHUMP00000011225; OTTHUMP00000011226; ple; Protein Pale; TH; The; TY3H_HUMAN; TYH; Tyrosine 3 hydroxylase; Tyrosine 3 monooxygenase; Tyrosine 3-hydroxylase; Tyrosine 3-monooxygenase; Tyrosine hydroxylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9282