Learn More
Biomatik Corporation Human Pulmonary surfactant-associated protein D, SP-D ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40775
Encompass_Preferred
The Human Pulmonary surfactant-associated protein D, SP-D ELISA Kit allows for quantitative determination of surfactant protein D in serum, plasma, bronchoalveolar lavage fluid and tissue homogenates. Species Reactivity: Human. Detection Range: 1.25 ng/mL-80 ng/mL. Sensitivity: 0.31 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P35247. Restriction: For Research Use Only.
Catalog No. 50-149-9313