Learn More
Biomatik Corporation Monkey Progesterone (PROG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC40989
Encompass_Preferred
Monkey Progesterone (Prog) Elisa Kit for detecting Progesterone(Prog) in serum, plasma. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 0.4-30ng/ml. Sensitivity: 0.2ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9402