Learn More
Biomatik Corporation Human Apolipoprotein A2, apo-A2 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41023
Encompass_Preferred
The Human Apolipoprotein A2, apo-A2 ELISA Kit allows for quantitative determination of apolipoprotein A-II in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 1 ng /mL-800 ng/mL. Sensitivity: 1 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P02652. Restriction: For Research Use Only.
Catalog No. 50-149-9415