Learn More
Biomatik Corporation Human Angiotensin 1-7 (Ang1-7) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41131
Encompass_Preferred
The Human Angiotensin 1-7 (Ang1-7) ELISA Kit allows for quantitative determination of Angiotensin 1-7 (Ang1-7 in serum, plasma, cell culture supernates. Species Reactivity: Human. Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.95 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-9455