Learn More
Biomatik Corporation Human Folic acid (FA) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKC41262
The Human Folic acid (FA) ELISA Kit allows for quantitative determination of Folic acid (FA) in serum, plasma. Species Reactivity: Human. Detection Range: 12.5 pg/mL-200 pg/mL. Sensitivity: 6.25 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.