Learn More
Biomatik Corporation Human Allograft inflammatory factor 1 (AIF1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41348
Encompass_Preferred
The Human Allograft inflammatory factor 1 (AIF1) ELISA Kit allows for quantitative determination of allograft inflammatory factor 1 in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.9 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P55008. Restriction: For Research Use Only.
Catalog No. 50-149-9510