Learn More
Biomatik Corporation Mouse CD63 antigen (CD63) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41424
Encompass_Preferred
Mouse Cd63 Antigen (Cd63) Elisa Kit for detecting Cd63 Molecule in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P41731; Cd63. Target Synonyms: Cd63CD63 antigen; CD antigen CD63. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9524