Learn More
Biomatik Corporation Mouse Cathepsin S (CTSS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41499
Encompass_Preferred
Mouse Cathepsin S (Ctss) Elisa Kit for detecting Cathepsin S in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O70370; Ctss. Target Synonyms: Ctss; CatsCathepsin S; EC 3.4.22.27. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9538