Learn More
Biomatik Corporation Human Fibronectin type III domain-containing protein 5 (FNDC5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41584
Encompass_Preferred
The Human Fibronectin type III domain-containing protein 5 (FNDC5) ELISA Kit allows for quantitative determination of fibronectin type III domain containing 5 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human. Detection Range: 7.8 ng/mL-500 ng/mL. Sensitivity: 1.95 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q8NAU1. Restriction: For Research Use Only.
Catalog No. 50-149-9560