Learn More
Biomatik Corporation Rat heparin-binding EGF-like growth factor (HBEGF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41619
Encompass_Preferred
Rat Heparin-Binding Egf-Like Growth Factor (Hbegf) Elisa Kit for detecting Heparin-Binding Egf-Like Growth Factor in serum, plasma, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q06175; Hbegf. Target Synonyms: Hbegf; Dtr; Hegfl; Proheparin-binding EGF-like growth factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9571