Learn More
Biomatik Corporation Human Lysyl oxidase homolog 2 (LOXL2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41760
Encompass_Preferred
The Human Lysyl oxidase homolog 2 (LOXL2) ELISA Kit allows for quantitative determination of lysyl oxidase-like 2 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human. Detection Range: 37.5 pg/mL-2400 pg/mL. Sensitivity: 9.375 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9Y4K0. Restriction: For Research Use Only.
Catalog No. 50-149-9588