Learn More
Biomatik Corporation Mouse Semaphorin-3A (SEMA3A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC41905
Encompass_Preferred
Mouse Semaphorin-3A (Sema3A) Elisa Kit for detecting Sema Domain, Immunoglobulin Domain (Ig), Short Basic Domain, Secreted, (Semaphorin) 3A in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O08665; Sema3a. Target Synonyms: Sema3a; Semad; SemD; Semaphorin-3A; Semaphorin III; Sema III; Semaphorin-D; Sema D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-9613