Learn More
Biomatik Corporation Mouse Protein Wnt-16 (WNT16) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKC42020
Mouse Protein Wnt-16 (Wnt16) Elisa Kit for detecting Wingless-Type Mmtv Integration Site Family, Member 16 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9QYS1; Wnt16. Target Synonyms: Wnt16; Protein Wnt-16. Restrictions: For Research Use Only. Not for use in diagnostic procedures.