Learn More
Biomatik Corporation Human AMBP (Alpha-1-Microglobulin/Bikunin Precursor) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE60234
Encompass_Preferred
The Human AMBP (Alpha-1-Microglobulin/Bikunin Precursor) ELISA Kit allows for quantitative determination in Serum, Plasma, Biological Fluids. Species Reactivity: Human. Detection Range: 4.69-300 ng/mL. Sensitivity: 2.81 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich. Detection Method: Colorimetric. Assay Time: 4.5 hours. UniProt ID: P02760. Cross Activity: No significant cross-reactivity or interference was observed. Restriction: For Research Use Only.
Numéro de catalogue. 50-149-9830
Il en reste null
ajouter au panier
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.