Learn More
Biomatik Corporation Human TDP43 (TAR DNA Binding Protein 43) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE60605
Encompass_Preferred
Human TDP43 (TAR DNA Binding Protein 43) ELISA Kit for detecting TDP43 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.31-20ng/ml. Sensitivity: 0.19ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: Q13148. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0082