Learn More
Biomatik Corporation Human TGF-2 (Transforming Growth Factor Beta 2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE60957
Encompass_Preferred
Human TGF-β2 (Transforming Growth Factor Beta 2) ELISA Kit for detecting TGF-β2 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P61812. Target Synonyms: BSC-1 cell growth inhibitor; Cetermin; Glioblastoma-derived T-cell suppressor factor; G-TSF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0314