Learn More
Biomatik Corporation Human VASPIN (Visceral Adipose Specific Serine Protease Inhibitor) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE61045
Encompass_Preferred
Human VASPIN (Visceral Adipose Specific Serine Protease Inhibitor) ELISA Kit for detecting VASPIN in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: Q8IW75. Target Synonyms: SERPINA12; OL-64; Visceral Adipose Tissue-Derived Serine Protease Inhibitor; Serine Proteinase Inhibitor; Clade A Member 12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0372