Learn More
Biomatik Corporation Human REG1 (Regenerating Islet Derived Protein 1 Alpha) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE61235
Encompass_Preferred
Human REG1α (Regenerating Islet Derived Protein 1 Alpha) ELISA Kit for detecting REG1α in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P05451. Target Synonyms: REG1A; ICRF; P19; PSP; PSPS; PSPS1; PTP; REG; MGC12447; Pancreatic Stone Protein; Pancreatic Thread Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0497