Learn More
Biomatik Corporation Human TFR1 (Transferrin Receptor 1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKE61281
Encompass_Preferred
Human TFR1 (Transferrin Receptor 1) ELISA Kit for detecting TFR1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.94ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colormetric. Assay Time: 3h, 30min. Accession Number: P02786. Target Synonyms: TfR1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-0532