Learn More
Biomatik Corporation Human ELN (Elastin) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKF57343
Encompass_Preferred
The Human ELN (Elastin) ELISA Kit allows for quantitative determination of ELN in Serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.469-30 ng/mL. Sensitivity: 0.281 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Sandwich ELISA, Double Antibody. UniProt ID: P15502. Restriction: For Research Use Only.
Catalog No. 50-150-0865